Pipetten met graduatie, Type 3, Volac®
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R400/A/5 R400/C/5 R400/D/5
612-1606EA 18.5 EUR
612-1606 612-1608 612-1609
Pipetten met graduatie, Type 3, Volac®
Pipetten Gegradueerde pipetten
Glass, zero at top, capacity at outlet.
  • Amber inscriptions
  • Sizes 5 ml and above are constricted for cotton plugging
  • Colour coded
Certificaten: BS 700