Pipette controller, Pipet-Aid® XP
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4-000-202 4-000-201-E 4-000-102 4-000-101
613-4253EA 1430 EUR
613-4253 DRUM4-000-201-E DRUM4-000-102 DRUM4-000-101
Pipette controller, Pipet-Aid® XP
Pipetten Pipetvullers
This lightweight, compact controller is specifically designed to minimize operator fatigue. It accepts most pipettes from 1 to 100 ml.
  • Three-speed settings provide more precise control for both aspirate and dispense modes
  • Easy access battery (9 V (Ni-MH)) replacement
  • EZ Grip nosepiece with double membrane filter
  • Small and lightweight
Each aspiration and dispense button has three speeds, (F) Fast, (M) Medium, (S) Slow. The medium speed is suitable for most routine techniques. Fast speed can deliver up to 50 ml in 5 seconds and can be used for rapidMore Product Information
Order Now
Specificaties
Environmentally Preferable
Type accu's 9V NiMH
Gewicht 78/9 oz.