Pipetteerballonnen
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4200003 4100003 4200005 4250003
612-9952EA 13.8 EUR
612-9952 612-9953 DENE4200005 DENE4250003
Pipetteerballonnen
Pipetten Pipetvullerballonnen
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).