Sterilisation Boxes for Pipettes
Leverancier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A20 A40 S20 S40 A30 A45 S30 S45
612-2085EA 37.3 EUR
612-2085 612-2086 612-2087 612-2088 612-2089 612-2090 612-2091 612-2092
Sterilisation Boxes for Pipettes
Dozen Pipetsterilisatiecontainers
GLW Pipette Boxes are made from aluminum. Due to its square-shaped form, the box remains stationary while working and will not roll away as is the case with round containers. The box is available in four different sizes.
  • Can be autoclaved at +121 °C; can withstand temperatures up to +250 °C
  • 'Economy' boxes: without silicone
  • 'Premium' boxes: integral silicone coating in base and lid
  • W×D (mm): 68×68
W×D (mm): 68×68