Reagentiareservoirs
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9510027 9510037 9510047
613-2671EA 18.3 EUR
613-2671 613-2672 612-6572
Reagentiareservoirs
Reagensreservoirs
Wit PP, autoclaveerbaar.
  • V-bodem
  • Voor 4-, 8- of 12-kanaalspipetten