Pasteurpipetten, glas
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M4150NO250SP4 M4230NO250SP4
612-3813EA 27.8 EUR
612-3813 612-3814
Pasteurpipetten, glas
Pipetten Pasteur-/transferpipetten
Pasteur pipette with a narrow tip. The suction tube is designed for cotton plugging.
Dispensing box with 250 pipettes.