Rekken voor vials, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
868805 985800
212-6191EA 300 EUR
212-6191 212-6192
Rekken voor vials, WHEATON®
Rekken Rekken voor chromatografievials
These polypropylene racks accommodate 36 or 50 headspace auto-sampler vials. Each well has an opening in the bottom to facilitate drainage.
  • Autoclavable and stackable
  • Chemically resistant
  • Include position identifiers