Rekken voor microbuisjes
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8760 8770-11 8780
700-2830EA 41.5 EUR
700-2830 700-2831 700-2832
Rekken voor microbuisjes
Rekken Tube Racks
PP, gekleurd met deksel.
  • Perfect for freezer use
  • Side handles for secure handling, snap-in-place lid
  • Numbered for easy sample identification
For 96 microtubes with 0,5 on one side or or 1,5 ml capacity on the other side. Arrangement of 8×12, conical tapered openings to hold tubes securely.