Antibèta-afvaldoos voor grote containers, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6745-0010 6745-9024
113-8170EA 789 EUR
113-8170 113-2809
Antibèta-afvaldoos voor grote containers, Nalgene®
Schermen Stralingsschermen
Acryl, transparant, dikte: 9 mm.
  • Large hinged door on cover allows easy access to the waste receptacle
  • Leakproof tray contains spills
  • Hinged corners allow removal of receptacle
Completely encloses a large container holding solid or liquid radioactive waste. Accommodates 20 litre carboys and standard office size trash cans. Smaller bin accommodates 10 litre containers.
W×D×H (inner): 406×406×736 mm (20 litre) 286×286×406 mm (10 litre).