Deksel vlakke flens met vier of vijf halzen, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAF4/41 MAF2/32
201-8062EA 249 EUR
201-8062 201-8579
Deksel vlakke flens met vier of vijf halzen, Quickfit®
Deksels Deksels voor reactievaten
Lids, PYREX® borosilicate glass, with precision ground, flat flange joints.
  • For use with Quickfit® reaction flasks and culture vessels
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
Specificaties
Environmentally Preferable