Scheppen
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9030 9010 9020 9040
NICK9030EA 78.8 EUR
NICK9030 231-0350 231-0351 231-0352
Scheppen
Scheppen
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
  • Excellent thermal resistance, autoclavable
  • Non magnetic, bright finish
  • Highly polished surface makes cleaning easier