Zelfklevende PCR-films en folies
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T329-1 T329-2 T329-5
293-1401EA 122 EUR
293-1401 293-1402 293-1405
Zelfklevende PCR-films en folies
Afdichtingen Afdichtingen van microtiterplaten
Transparante zelfklevende afdichtingsfilm is ideaal voor het verminderen van kruisbesmetting en/of morsen bij PCR-toepassingen. 
  • Thermostable and functional from −70 to +95 °C
  • Choice of sterile or non sterile
Aluminium sealing foil is suitable for both manual sealing during PCR work and high throughput applications. Pierceable with a pipette tip for easy access to sample. Adhesive backing makes it easy to apply. Will resist temperatures from –80 to +120 °C. It is recommended that the foil be applied using the specially designed rubber roller toMore Product Information