Lepel spatel, brede vorm
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2003 2004 2001 2002
231-0076EA 38.9 EUR
231-0076 231-0077 231-0156 231-0075
Lepel spatel, brede vorm
Spatels
Pure nickel (99,6%).
  • One end is flat and spatula-shaped, the other end is spoon-shaped
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean