Dubbelspatel, Chattaway, nikkel
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2202 2211 2213 2215 2214 2203 2210 2216
NICK2202EA 71.4 EUR
NICK2202 231-0160 231-0162 231-0164 231-0163 231-0207 231-0209 NICK2216
Dubbelspatel, Chattaway, nikkel
Spatels
Multipurpose spatula made of pure nickel (99,6%) for a wide range of uses. One end is flat and spatula-shaped, the other end is rounded and turned up at 45°. Available in light or medium weight design.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean