Dubbelspatel
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3100G 3101G 3102G 3103G 3104G 3105G
231-2121EA 4.1 EUR
231-2121 231-2122 231-2123 231-2124 231-2125 231-2126
Dubbelspatel
Spatels
Stainless steel 18/10, anti-magnetic. Multi-purpose spatula for a wide range of uses.
  • With round handle, one flat and one rounded end
  • Curved shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance