Sampling spoons with long handle, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F367270000 F367230000 F367240000 F367260000 F367250000
231-0005EA 67.6 EUR
231-0005 231-0366 231-0367 231-0368 231-0365
Sampling spoons with long handle, SP Bel-Art
Lepels
Designed to gather small amounts of sample material, these PP sampling spoons are available in four bowl sizes.
  • Long handle reaches easily into jars, bottles and other confined areas; minimises the potential of personal contact with sampled material
  • FDA grade materials; safe for food, drug and cosmetic contact
* assortiment met telkens 3 van bovenstaande lepels.