Water estimators, Dean-Stark, light entrainers, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WE6/23/25 WE6/23/2 WE6/23/7 WE6/23/10T
201-0259EA 130 EUR
201-0259 201-8752 201-8754 201-8751
Water estimators, Dean-Stark, light entrainers, Quickfit®
Onderdelen voor destillatie Destillatiesets
PYREX®, borosilicate glass.
  • For use with solvent entrainers which are lighter than water
  • Graduated tube, item 201-8751 has stopcock for easy drainage
Dean & Stark pattern receivers for the determination of the water content in petroleum products.
BS 756
Certificaten: BS 756