Plugkraan
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SST104 SST102 SST203 SST103 SST106
201-0569EA 168 EUR
201-0569 BIBBSST102 BIBBSST203 BIBBSST103 BIBBSST106
Plugkraan
Afsluitkranen
PYREX®, borosilicate glass with PTFE key.
  • Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
  • Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
  • Made to a taper of 1:10 giving a combination of performance and ease of operation