Driewegskraantjes, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6470-0002 6470-0004
216-4424EA 63.3 EUR
216-4424 216-4425
Driewegskraantjes, Nalgene®
Afsluitkranen
PP, translucent with PTFE tap, white.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Non blocking, sealed tap, no lubricant required
Taps toelopende driewegkranen in T-opstelling
For regulating or blocking the flow of liquids in tube connections. For tubing with internal diameter 6,4 mm to 9,5 mm.