Adapter, kegel met kraan, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF11/2/SC MF11/3/SC MF11/4/SC MF11/1/SC
201-8596EA 91.7 EUR
201-8596 201-8599 201-3009 201-0263
Adapter, kegel met kraan, Quickfit®
Adapters Adapters, Slijpstukken met slangaansluitingen
Adapter, PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
General purpose (GP) Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screw thread connectors allow rubber hose etc. to be fitted easily and safely.