Adapter met kraan, T vorm met kegel, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF11/1B/SC MF11/2B/SC MF11/3B/SC
201-0264EA 105 EUR
201-0264 201-0265 201-0266
Adapter met kraan, T vorm met kegel, Quickfit®
Adapters Adapters, Slijpstukken met slangaansluitingen
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
General purpose (GP) Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber tubing etc. to be fitted easily and safely.