Push-in stoppen voor buizen
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T401-10N T401-3DSPE T401-10W T401-10B T401-10G T401-10R T401-10Y T401-10DSPP T401-3DSPP T401-3N T401-3B T401-3G T401-3R T401-3W T401-3Y
212-9606EA 97.2 EUR
212-9606 SIMPT401-3DSPE SIMPT401-10W SIMPT401-10B SIMPT401-10G SIMPT401-10R SIMPT401-10Y SIMPT401-10DSPP SIMPT401-3DSPP SIMPT401-3N SIMPT401-3B SIMPT401-3G SIMPT401-3R SIMPT401-3W SIMPT401-3Y
Push-in stoppen voor buizen
Stoppen PE/LDPE/HDPE stoppen
HDPE. Fit tubes of 15 ml capacity and 17 mm Ø.
  • Impermeable to liquids
  • Can be inserted in tubes and removed again easily with one hand
  • Economical, can also be used as disposable stoppers