Conische rubberen stoppen, stevig
Leverancier: LEY RUBBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB003/S SB004/S SB005/S SB006/S SB007/S SB008/S SB009/S SB010/S SB011/S SB013/S SB015/S SB017/S SB019/S SB021/S SB023/S SB025/S SB027/S
LEYRSB003/SEA 53.2 EUR
LEYRSB003/S LEYRSB004/S LEYRSB005/S LEYRSB006/S LEYRSB007/S LEYRSB008/S LEYRSB009/S LEYRSB010/S LEYRSB011/S LEYRSB013/S LEYRSB015/S LEYRSB017/S LEYRSB019/S LEYRSB021/S LEYRSB023/S LEYRSB025/S LEYRSB027/S
Conische rubberen stoppen, stevig
Stoppen Siliconen stoppen
These solid stoppers are made from translucent silicone rubber.
  • Autoclavable to 121 °C
  • Wide temperature resistance from −60 to +200 °C
  • High chemical and electrical resistance
Certificaten: Conforms to BS 2775
Order Now
Specificaties
Environmentally Preferable