Rode rubberen stoppen
Leverancier: LEY RUBBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RB023/1H RB021/1H RB013/1H RB015/1H
LEYRRB023/1HEA 36 EUR
LEYRRB023/1H LEYRRB021/1H LEYRRB013/1H LEYRRB015/1H
Rode rubberen stoppen
Stoppen Rubberen stoppen
Red rubber stoppers as Cat. No. 391-2100 but with one standard hole of 5 mm diameter.