Conische kurken stoppen
Leverancier: IBS Stock Items
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CK0705 CK0907 CK1411 CK1613 CK1815 CK2017 CK2118 CK2320 CK3126 CK3329 CK3828 CK3834 CK4037 CK4238 CK4540 CK5045 CK5550 CK10095 CK105100 CK110105 CK115110 CK120115 CK125120 CK130125 CK135130 CK140135 CK6055 CK6560 CK7065 CK7570 CK8075 CK8580 CK9085 CK9590 CK145140 CK150145
217-0598EA 0 EUR
217-0598 217-0600 217-0614 217-0618 217-0620 217-0622 217-0623 217-0625 217-0630 217-0633 217-0635 217-0636 217-0637 217-0638 217-0639 217-0640 217-0641 217-0602 217-0603 217-0604 217-0606 217-0607 217-0609 217-0610 217-0612 217-0613 217-0642 217-0643 217-0644 217-0645 217-0646 217-0647 217-0648 217-0649 217-0615 217-0616
Conische kurken stoppen
Stoppen Kurkstoppen
Natural cork A1 quality, low porosity.
  • Light, good fit
  • Holes can be made easily with a cork drill, or the stoppers can be cut to size
  • Convenient pack sizes
Order Now
Specificaties
Environmentally Preferable