NS-stoppen
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01400512 01400529 01400514 01400519
217-9096EA 9.8 EUR
217-9096 217-9099 217-9097 217-9098
NS-stoppen
Stoppen Glazen stoppen
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
  • Excellent chemical resistance
  • High thermal resistance and minimal heat expansion results in high resistance to temperature changes
  • Octagonal top allows easy release and removal of the stopper
Certificaten: DIN 12252