NS-stoppen
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01400007 01400010 01400012 01400014 01400019 01400024 01400029 01400034 01400045 01400345 01400307 01400324 01400314 01400312 01400329 01400310 01400319 01400334
217-9101EA 3.3 EUR
217-9101 217-9102 217-9103 217-9105 217-9106 217-9108 217-9110 217-9112 217-9114 LENZ01400345 LENZ01400307 217-9095 217-9093 LENZ01400312 LENZ01400329 LENZ01400310 217-9094 217-0763
NS-stoppen
Stoppen Glazen stoppen
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certificaten: DIN 12252
Bestelinformatie: Available with flat or pointed bottom.