Absorberend papier, BenchGuard® en BenchGuard® Extra, Sterilin®
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BG50E BG50 BG60E
115-9321EA 331 EUR
115-9321 115-9305 115-9320
Absorberend papier, BenchGuard® en BenchGuard® Extra, Sterilin®
Oppervlakbescherming Werkbladbescherming
Highly absorbent paper for protecting benches and surfaces against liquid spills.
  • One side plastic coated to prevent soak through
  • Available in either roll or sheet forms
  • BenchGuard Extra®: double absorbency for more demanding applications
Absorption of 400 ml/m² for BenchGuard® and 800 ml/m² for BenchGuard® Extra.
Leveringsinformatie: Supplied in easy to use dispenser packs.