Tafelbeschermers, Benchkote®, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2300-599 2300-731 2300-772 2300-916 2300-594 2300-917
115-9204EA 132 EUR
115-9204 115-9201 115-9202 115-9203 115-9209 115-0588
Tafelbeschermers, Benchkote®, Whatman™
Oppervlakbescherming Werkbladbescherming
Cytiva's Whatman™ Benchkote® surface protector is an absorbent lab paper that protects surfaces against hazardous spills. After use, the bench protector sheet can be incinerated or disposed of according to local regulations.
  • Material is strong and tear resistant when wet or dry
  • Smooth white surface can be written on with ink or pencil and lies flat
  • Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Can be incinerated after use: PE layer is rapidly consumed when incinerated and does not melt or drip
Benchkote® material has a high quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.