Sponzen wissers, Polywipe®
Leverancier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MW728 MW729
720-0144EA 160 EUR
720-0144 720-0145
Sponzen wissers, Polywipe®
Wattenstaafjes Wattenstaafjes voor monstername
Non-inhibitory sponge material allows effective sampling of larger areas for environmental and hygiene monitoring. Available dry or pre-moistened with sterile diluent. Polywipes are supplied in peel pouches for immediate use, or in a 110 ml blue-capped container for transfer to the laboratory.
  • Blue colour for visibility
  • Triple wrapped and radiation sterilised
Leveringsinformatie: Supplied with individual sterile glove for handling.