Sampling swabs, Dryswab™
Leverancier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MW108 MW108/5 MW112 MW1048 MW1028
720-0127EA 301 EUR
720-0127 720-0128 720-0129 720-2203 MEDWMW1028
Sampling swabs, Dryswab™
Wattenstaafjes Wattenstaafjes voor monstername
These dry swabs are ideal for collecting specimens or samples for analysis or for transferring materials during a diagnostic procedure.
  • Sterile, ready-to-use
  • Available with different shaft and head materials
  • Rayon swabs are particularly suitable for the recovery of microorganisms
Labelled tubes: Each swab comes self-contained and ready to use with its own tube for easy transport to the laboratory. The swab is mounted in the tube plug which serves as a convenient holder so there is no need for direct handling of the swab. Each tube and cap assembly isMore Product Information