Standaard microliterspuiten
Leverancier: Hospidex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EX-MS250 EX-MS500
613-1719EA 75.7 EUR
613-1719 613-1720
Standaard microliterspuiten
Spuiten Injectiespuiten voor chromatografie
Providing ultra-precise analyses. The plunger extends down into the tip of the needle ensuring that no dead volume is created.
The bevelled needle tip is 50 mm long.