HPLC-spuiten, Rheodyne
Leverancier: Hospidex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EX-MSR05 EX-MSR10 EX-MSR25 EX-MSR50 EX-MSR250 EX-MSR500
613-1728EA 72.7 EUR
613-1728 613-1729 613-1730 613-1731 613-1733 613-1734
HPLC-spuiten, Rheodyne
Spuiten Injectiespuiten voor chromatografie
For ultra-precise analyses. The plunger extends down into the tip of the needle ensuring that no dead volume is created.
Model with interchangeable needle on request. HPLC syringes for Waters and Valco on request.