Injectienaalden
Leverancier: ROMETSCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13201-100100 13201-20
612-0301EA 114 EUR
612-0301 612-0303
Injectienaalden
Injectienaalden
Rust-proof, reusable injection needles with nickel-plated Luer lock fitting.
  • Cannulas for puncturing and injecting
  • Normal bevelled tip