Tapes, construction and maintenance
Leverancier: ULTRATAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1160WH100-P3DU 1160WH200-P3DU 1160BK260-P3DU 1160YE100-P3D 1160RE260-P3D 1160BL260-P3D 1160RE075-P3D 1160BL200-P3DU 1160YE200-P3DU 1160GR100-P3D 1160WH050P3D 1160GR200-P3DU 1160RE200-P3D 1160BL200-P3G 1160YE260-P3D 1160BL100-P3DU 1160OR100-P3D 1160RE100-P3DU
817-0070EA 189 EUR
817-0070 817-0071 UTAP1160BK260-P3DU UTAP1160YE100-P3D UTAP1160RE260-P3D UTAP1160BL260-P3D UTAP1160RE075-P3D UTAP1160BL200-P3DU UTAP1160YE200-P3DU UTAP1160GR100-P3D UTAP1160WH050P3D UTAP1160GR200-P3DU UTAP1160RE200-P3D UTAP1160BL200-P3G UTAP1160YE260-P3D UTAP1160BL100-P3DU UTAP1160OR100-P3D UTAP1160RE100-P3DU
Tapes, construction and maintenance
Tape
These cleanroom tapes are made of vinyl with rubber adhesive and are suitable for construction and maintenance.
  • Combines particle-free cleanliness with the ability to adhere to a variety of materials, including plastic and metal
  • Abrasion and moisture resistant and unaffected by most chemicals
  • Performs well over a wide temperature range
  • Can be written on by cleanroom pen
Suitable for use in ISO Class 4 environments.
Length: 32,9 m
Verschillende kleuren beschikbaar.
Verpakking: Gereinigd en dubbel verpakt onder cleanroom-omstandigheden.