Packaging tape, wafer box tape
Leverancier: ULTRATAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1110CL100-P3T 1110WH100-P3T 1110WH200-P3T 1110BL075-P3T 1110CL200-P3T 1110YE075-P3TU 1110BL100-P3T 1110RE050-P3T 1110RE100-P3T 1110BL200-P3T
817-1710EA 0 EUR
817-1710 817-0068 UTAP1110WH200-P3T UTAP1110BL075-P3T UTAP1110CL200-P3T UTAP1110YE075-P3TU UTAP1110BL100-P3T UTAP1110RE050-P3T UTAP1110RE100-P3T UTAP1110BL200-P3T
Packaging tape, wafer box tape
Tape
These PE tapes are designed specifically for sealing wafer boxes. They are also used for sealing plastic bags and for other cleanroom uses.
  • Adheres lightly to plastics, stretching easily around corners
  • Low metal content
  • No residues after prolonged storage
  • Easy release from the roll with low static charge
Length: 33,0 m.
Different colours available.
Verpakking: Supplied clean and double bagged.