Cleanroomtapes, Supertack™
Leverancier: ULTRATAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1153WH100-P3DU 1153BW200-P3D 0153BW200-P3S 0153BW100-P3S 1153YE200-P3D 1153WH600-P3D 1153YE600-P3D 1153RE600-P3D 1153RE100P3D_U 1153YE100P3D 1153BL600-P3D 1153GR600-P3D 1153OR600-P3D 1153BW600-P3D 1153BW100-P3D 1153WH300-P3D 0153BL050-P3S 0153BL200-P3S
UTAP1153WH100-P3DUEA 28.6 EUR
UTAP1153WH100-P3DU UTAP1153BW200-P3D UTAP0153BW200-P3S UTAP0153BW100-P3S UTAP1153YE200-P3D UTAP1153WH600-P3D UTAP1153YE600-P3D UTAP1153RE600-P3D UTAP1153RE100P3D_U UTAP1153YE100P3D UTAP1153BL600-P3D UTAP1153GR600-P3D UTAP1153OR600-P3D UTAP1153BW600-P3D UTAP1153BW100-P3D UTAP1153WH300-P3D UTAP0153BL050-P3S UTAP0153BL200-P3S
Cleanroomtapes, Supertack™
Tape
These Supertack™ cleanroom tapes are made of PE with rubber adhesive and are ideal for any application that requires a strong bond.
  • Tapes remove cleanly with no residue
  • Tear by hand
  • Accept cleanroom ballpoint pen writing
The tape is available in Protocol™ uniquely striped color pattern which allows immediate recognition of the product as a certified cleanroom tape. Suitable for use in ISO Class 4 environments
Lengte: 32,9 m.
Verschillende kleuren beschikbaar.
Verpakking: gereinigd en dubbel verpakt onder cleanroom-omstandigheden.