Milieuvriendelijke thermometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G11374-EF G11376-EF G11378-EF G11380-EF G11382-EF G11388-EF G11392-EF G11396-EF G11404-EF G11392-EF/DKD3
AMARG11374-EFEA 30.8 EUR
AMARG11374-EF AMARG11376-EF AMARG11378-EF AMARG11380-EF AMARG11382-EF AMARG11388-EF AMARG11392-EF AMARG11396-EF AMARG11404-EF AMARG11392-EF/DKD3
Milieuvriendelijke thermometers
Thermometers Analoge thermometers
These thermometers are designed for everyday use thanks to a non toxic green fluid. Choose either solid stem to reduce risk of accidental breakage or enclosed scale with milk glass for additional accuracy.
  • Veilig
  • Kwikvrij
  • Robuuste schacht of omsloten schaal
Vulling : Organische vloeistof
Length: 300 mm
Immersion depth: 76 mm