Tangen voor smeltkroezen, gebogen
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3060 3061 3063 3064 3066
NICK3060EA 14.6 EUR
NICK3060 231-0357 NICK3063 NICK3064 231-0358
Tangen voor smeltkroezen, gebogen
Tangen
Stainless steel 18/10, anti-magnetic. For easy, safe handling of crucibles made from a wide variety of materials.
  • With grooved tips
  • Excellent corrosion resistance
  • Good thermal and electrical conductivity
  • Excellent thermal resistance