Sterilisatiebakken, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6900-0010 6900-0020
489-2120EA 57.7 EUR
489-2120 489-2121
Sterilisatiebakken, Nalgene®
Reservoirs/Schalen/Potten Trays/pannen voor sterilisatie
PP.
  • Rolled edges for extra stiffness
  • Handles for easy carrying
For drying and sterilising instruments and labware, collecting and washing soiled lab equipment.