Driepikkels
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5131 5132 5133 5134 5141 5142 5143 5135 5145 5136 5146 5144
451-0092EA 16.6 EUR
451-0092 451-0093 451-0094 451-0095 451-9101 451-9102 451-9103 451-0157 451-0160 451-0158 451-0161 451-0159
Driepikkels
Driepikkels
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.