Utility carts, MW series
Leverancier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MW703 MW705 MW711 MW104 MW103 MW401 MW402 MW403 MW404 MW406 MW105 MW203 MW702 MW208 MW206 MW604 MW106 MW108 MW204 MW205 MW601 MW603 MW605 MW606 MW607 MW608 MW611 MW612 MW701 MW704 MW706 MW707 MW708 MW602 MW712
139-9995EA 601 EUR
139-9995 139-9996 139-9997 139-0777 139-0776 139-0077 139-0078 139-0079 139-0080 139-0081 139-0778 139-0372 139-0388 139-0781 139-0375 139-0377 139-0779 139-0780 139-0373 139-0374 139-0381 139-0382 139-0383 139-0378 139-0384 139-0379 139-0385 139-0380 139-0386 139-0389 139-0390 139-0387 139-0391 139-0376 139-0392
Utility carts, MW series
Transportwagens
These steel or chrome plated carts consist of Super Erecta shelves with plastic split sleeves, two one-piece handles of matching finish, and designated castors with donut bumpers. These multipurpose trolleys are perfect for carrying out an enormous variety of tasks.
  • Ergonomically designed for low effort transport
  • Precise handling for manoeuvring in close quarters
  • Sturdy aluminum corner castings firmly secure solid shelves to posts
  • Stable, tough, simple and easy to clean
  • MW carts use light-duty resilient castors