Centrifuge tubes
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
120180_2000 187262_1000 191180_1000 115262_2000
212-3166EA 304 EUR
212-3166 391-3476 525-0406 525-0405
Centrifuge tubes
Buisjes Centrifugebuisjes
Transparent PS or PP, 2-position ventilation stopper, round bottom, sterile.
  • PS tubes without graduations, good thermal, mechanical and chemical resistance, centrifugation RFC 5000×g (fix-angle rotor) or 4800×g (swinging-bucket rotor)
  • PP tubes with graduations; high thermal, mechanical and chemical resistance, centrifugation RCF 34000×g (fix-angle rotor) or 5800×g (swinging-bucket rotor)
  • The ventilation stopper has two functions: when pushed in gently, it provides uniform ventilation of the tube contents; when pushed in firmly, it provides a tight seal