Centrifugebuizen, ASTM, BLAUBRAND®
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
362038 362138 362338
525-1272EA 41.2 EUR
525-1272 525-1273 525-1271
Centrifugebuizen, ASTM, BLAUBRAND®
Buisjes Centrifugebuisjes
Borosilicaatglas 3.3, met graduatie.
  • Withstands forces up to 700×g
  • Design, accuracy etc., to meet ASTM specifications
  • For water and sediments in crude oils according to ASTM D91/D96