Centrifugeerbuisjes
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
778012 779012 779022
525-1242EA 61 EUR
525-1242 525-0500 525-1005
Centrifugeerbuisjes
Buisjes Centrifugebuisjes
AR® glass, conical bottom, beaded rim (except 525-0500 cone-pointed bottom, rimless).
  • Bestand tot max. 3000×g