Schroefdoppen sluitingsset, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DS3131-0020 DS3131-0024 DS3131-0038
525-2913EA 59.1 EUR
525-2913 525-2914 525-2915
Schroefdoppen sluitingsset, Nalgene®
Sluitingen Reageerbuissluitingen
Tefzel™ FEP-plug, PP-schroefsluiting, Viton™ fluorelastomeer o-ring.
  • PP-schroefsluiting, ETFE-plug, Viton™ fluorelastomeer o-ring
  • Stevige sluiting voor gebruik op maximale snelheden zonder lekkage
  • Autoclaveerbaar/biohazard/lekvrij
Process tubes at their highest rated speed level (10000×g or greater) without leakage, even when full. Sealing cap assemblies include a Viton™ fluoroelastomer O-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.