Drukstoppen voor centrifugeerbuisjes, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DS3111-0016 DS3111-0017 DS3111-0018 DS3111-0025 DS3111-0029 DS3111-0030 DS3111-0032
525-2932EA 36.2 EUR
525-2932 525-2933 525-2934 525-2936 525-2937 525-2938 525-2939
Drukstoppen voor centrifugeerbuisjes, Nalgene®
Sluitingen Reageerbuissluitingen
PP, autoclaveerbaar.
  • Designed for both Nalgene® round bottom and conical centrifuge tubes
  • A tab facilitates the handling of the closure
Bestelinformatie: 525-2937 is for 50 ml tubes with rim and round bottom, 525-2938 is for 50 ml round bottom, rimless tubes.