Hollow-blown glass stoppers, C-length, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2595/01M 2595/02M 2595/03M 2595/05M 2595/06M 2595/07M
217-0734EA 137 EUR
217-0734 217-0735 217-0736 217-0737 525-0385 217-0738
Hollow-blown glass stoppers, C-length, Quickfit®
Stoppen Glazen stoppen
These glass stoppers are ideally suited for use with Quickfit® jointed glassware and other glassware having the shorter 'C' length ground glass sockets.
  • Chemical resistance
  • High temperature resistance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383.