Eppendorf® Protein LoBind® microcentrifugebuisjes
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030108.094 0030108.116 0030108.132 0030108.302 0030122.216 0030122.240 0030122.356 0030108.442 0030108.450 0030108.434 0030122.283
525-0132EA 16.4 EUR
525-0132 525-0133 525-0134 525-0792 EPPE0030122.216 EPPE0030122.240 EPPE0030122.356 EPPE0030108.442 EPPE0030108.450 EPPE0030108.434 EPPE0030122.283
Eppendorf® Protein LoBind® microcentrifugebuisjes
Buisjes Microcentrifugebuisjes
Engineered to minimise sample binding and elevate recovery, Eppendorf® Protein LoBind® tubes safeguard valuable proteins during processing. The cutting-edge LoBind® technology ensures optimal yields for Western blotting, mass spectrometry, and protein purification applications.
  • Bestand tot max. 18 000×g
  • Minimum protein verlies (minder dan 3%, BSA 1 µg/ml)
  • Bevat geen DNA, DNase, RNase en PCR-inhibitoren (PCR-vrij), oppervlak zonder coating, bijv. silicone
Unlock the benefits of Eppendorf® Protein LoBind® tubes, which are meticulously designed to offer unparalleled advantages. Available in a range of sizes and formats, these tubes are precision-crafted to minimise sample binding, providing a protective shield for valuable proteins, particularly in low-concentration applications. The LoBind® technology not only diminishes the risk of sample loss but also elevates protein recovery, guaranteeing users extract the maximum yield from every precious sample. Gain peace of mind knowing that proteins are safeguarded and results are optimised with Eppendorf® Protein LoBind® Tubes.
Applications include preparation or storage of protein, peptide, or antibody samples, Enzymatic assays (the hydrophilic surface reduces denaturation when it comes into contact with the tube inner wall), prevention of sample loss during storage of virus stock solutions, and storage of cell suspensions.
Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Material: PP
Sterility: Non sterile
Purity Grade: PCR clean