PCR microtubes with flat cap, Axygen®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PCR-02-B PCR-02-V PCR-02-Y
731-4000EA 68.96 EUR
731-4000 731-4004 731-4005
PCR microtubes with flat cap, Axygen®
Buisjes PCR-buisjes
Polypropylene tubes are specifically designed for PCR and thermal transfer applications.
  • Made from highly polished molds
  • Free of lubricants, dyes, heavy metals, and fillers
  • Certified RNase-, DNase-, and pyrogen-free to ensure the purity of your reaction
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling.